Zone Samplers for Powders and Granules
Supplier: SAMPLING SYSTEMS LTD
1610A-1000 1610A-1500 1610A-2000 1610A-2500 1610A-600
77576-298EA 233.66 USD
77576-298 77576-300 77576-302 77576-304 77576-306
Zone Samplers for Powders and Granules
Samplers Solid Samplers
An ideal tool for taking representative composite samples from containers, big bags or silos.
  • Aluminium construction
  • Takes multiple samples along its length
  • Easy to operate
The zone sampler is ideal for taking samples of powders and granules at multiple locations. The lightweight sampler is quick and easy to use.
The robust double tube design keeps material out of the sampler until it is opened.