Powder Triers
Supplier: SAMPLING SYSTEMS LTD
1216A-1500 1216A-1000 1216A-600
77576-066EA 1748.65 USD
77576-066 77576-064 77576-068
Powder Triers
Samplers Solid Samplers
These triers are suitable for taking a large volume, cross sectional sample from a container.
  • Robust all stainless steel construction with bright finish
  • Cutting scroll for the sampler to go into the product
  • Turn the handle to close the slot
Suitable for sampling cereals, grains, free flowing powders, granules and even slightly cohesive or compacted powders.
Certificaciones: GMP.