Immersion Cylinders for Liquid Sampling
Supplier: SAMPLING SYSTEMS LTD
5132B-500 5132S-1000 5132S-300 5132S-500 5132B-1000 5132B-300
77576-546EA 309.78 USD
77576-546 77576-548 77576-550 77576-552 77576-542 77576-544
Immersion Cylinders for Liquid Sampling
Samplers Fluid Samplers
These immersion cylinders are designed to take liquid samples from open water as well as tanks, drums, wells, and large storage containers. They are available made from either 304 stainless steel or brass, and are available in a range of sizes.
  • Stainless steel or brass construction
  • Available in 300, 500 and 1000 ml sizes
  • Ideal for either mid-point or bottom sampling
The Immersion Cylinder allows for two methods of sampling. When the cylinder reaches the bottom of the tank, it will automatically open. When the cylinder is mid-level, a second control line can be used to open the sampler.
These cylinders are designed to be easy to clean. Both ends of the sampler unscrew for convenient cleaning access.