SteriWare® Dosing Spoons, PS
Supplier: SAMPLING SYSTEMS LTD
8642A-01 8642A-01S
77577-420EA 45.22 USD
77577-420 77577-422
SteriWare® Dosing Spoons, PS
Spoons
These disposable and lightweight PS spoons are ideal for sampling and dosing small amounts of powder and liquid. Ideal for work in quality control, laboratories and production.
  • Made from white shatterproof high impact polystyrene (FDA and EU 10/2011 acceptable grade)
  • Handy design comes with a 2,5 and 5 ml spoon on either end
  • Available in both sterile and non sterile versions
  • Cross contamination is eliminated - the device can be used straight from the bag
  • Moulded and packed in a cleanroom to assure cleanliness and hygiene
  • BSE/TSE-free
Envase: Individually bagged and shipped with full documentation including certificates of conformity and sterilisation certificates.