SteriWare® Volumetric Spoons, Disposable
Supplier: SAMPLING SYSTEMS LTD
8640H-01 8640H-0123S 8640H-0246S 8640H-05 8640H-05S 8640H-10 8640H-10S 8640H-20 8640H-20S 8640H-01S
77577-400EA 90.96 USD
77577-400 77577-402 77577-406 77577-408 77577-410 77577-412 77577-414 77577-416 77577-418 77577-404
SteriWare® Volumetric Spoons, Disposable
Spoons
These volumetric spoons allow small volumes to be measured accurately. It made of HDPE and are designed to be used straight from the bag.
  • Moulded and packed in a cleanroom
  • BSE/TSE free
  • Full Batch Traceability
  • Fully validated sterilisation procedure. SAL 10⁻⁶
  • Available in a range of sizes from 1 to 20 ml
Suitable for food contact: FDA & EU 10/2011 conforming materials.
Envase: Individually bagged.