SteriWare® Powder Spatulas, Disposable, Cupped
Supplier: SAMPLING SYSTEMS LTD
8284B-150 8284B-150S 8284B-250 8284B-250S 8284R-150 8284R-150S 8284R-250 8284R-250S 8284A-150 8284A-150S 8284A-250 8284A-250S
77577-246EA 148.6 USD
77577-246 77577-248 77577-250 77577-252 77577-254 77577-256 77577-258 77577-260 77577-238 77577-240 77577-242 77577-244
SteriWare® Powder Spatulas, Disposable, Cupped
Spatulas
These cupped disposable powder spatulas are made of PS or PP. They are ideal for powder and granule sampling. The cupped design means that even very free flowing powders remain in the spatulas. The pointed tip also means it can pierce sacks and penetrate into dense powder.
  • Manufactured and packed in a controlled atmosphere (class 100000 cleanroom)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Suitable for use in ATEX Zones 0, 1 and 2
  • Available in different colours (red, blue, white)
  • Available sterile and non sterile
Ideal for production, quality checking and raw materials sampling. Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.).
Certificaciones: Materials conform to FDA, EU 10/2011 and EC 1935/2004. BSE/TSE free.
Envase: Individually wrapped.