Scoops, USP Class VI, Double-Bagged, ULTRA
Supplier: SAMPLING SYSTEMS LTD
9045H-1000S 9045H-125S 9045H-250S
77577-570EA 169.03 USD
77577-570 77577-572 77577-574
Scoops, USP Class VI, Double-Bagged, ULTRA
Scoops
The double bagged sterile scoops are made from high quality USP class VI HDPE. A box liner is also used giving a third layer of protection.
  • Double-bagged
  • Produced in Class 100,000 cleanroom, packed in Class 10,000 cleanroom
  • Sterilised by gamma irradiation
These scoops all have an ergonomic handle which is positioned high up the body so as to keep the hand as far away from the powder as possible.The scoops are designed to be used straight from the bag. The double bagging means that the outer bag can be removed whilst entering the cleanroom, ensuring the cleanliness inside the cleanroom or manufacturing area.
This range is ideal for sampling, production and dispensing small amounts.
Certificaciones: USP Class VI Medical grade polymer is FDA and EC compliant, tested to USP661.
Envase: Supplied in a lined box.