PharmaScoop® Scoops, Open, Stainless Steel
Supplier: SAMPLING SYSTEMS LTD
A336-1000 A336-1500 A336-2000 A336-250 A336-500 A336-750
77577-870EA 131.36 USD
77577-870 77577-872 77577-874 77577-876 77577-878 77577-880
PharmaScoop® Scoops, Open, Stainless Steel
Scoops
These general purpose scoops made from stainless steel 316 feature a heavy duty, crevice free construction which makes them ideal for use in the food and pharmaceutical industries.
  • Open top design, flat bottom with rounded edges
  • 316 stainless steel is a high-quality grade and is therefore very corrosion resistant
  • Satin finish