PharmaLabels Labels
Supplier: SAMPLING SYSTEMS LTD
8220P-003 8220P-004 8220P-005 8220P-01 8220P-02 8220P-03 8220P-04 8220P-05 8220P-06 8220P-001 8220P-002
77577-186EA 112.11 USD
77577-186 77577-188 77577-190 77577-192 77577-194 77577-196 77577-200 77577-202 77577-204 77577-182 77577-184
PharmaLabels Labels
Labels
PharmaLabels are a range of paper labels with a special adhesive that conforms to FDA and EC specifications.
  • Medium strength adhesion
  • Working temperature range –40 to +70 °C
PharmaLabels are ideal for applications where the adhesive might come into contact with the product.
Certificaciones: FDA acceptable adhesive conforms to FDA CFR175.105 and EC 1935/2004.