Ultra Adhesive Labels
Supplier: SAMPLING SYSTEMS LTD
8220A-001 8220A-002 8220A-003 8220A-004 8220A-005 8220A-006 8220A-01 8220A-02 8220A-03 8220A-04 8220A-05 8220A-06 8220A-30 8220A-31 8220A-32 8220A-33 8220A-34 8220A-35
77577-138EA 91.79 USD
77577-138 77577-140 77577-142 77577-144 77577-146 77577-148 77577-150 77577-152 77577-154 77577-156 77577-158 77577-160 77577-162 77577-164 77577-166 77577-168 77577-170 77577-172
Ultra Adhesive Labels
Labels
Ultra Adhesive (UA) Labels are special high quality labels with an ultra-adhesive foil backing.
  • High strength adhesive - will even stick on very dusty surfaces
  • Working temperature range –20 to +70 °C
  • Dust and weatherproof
UA Labels are ideal for sealing sacks after sampling. The label will not only record the fact that the sack has been sampled but it will prevent the sack from leaking powder as it is handled.