Wide Mouth Stainless Containers
Supplier: SAMPLING SYSTEMS LTD
A856-100 A856-50 A856-75
77578-142EA 1810.97 USD
77578-142 77578-144 77578-146
Wide Mouth Stainless Containers
Barrels, Kegs, Drums
The stainless steel wide mouth containers are perfect for multi purpose drums.
  • Suitable for manufacturing and storing powders, creams and liquids
  • Made from chemically resistant 316L stainless steel
  • Includes 6" tri ferrule lid system giving an excellent water and air tight seal
  • Mirror polish finish
  • Can be easily cleaned
These wide mouth stainless steel containers are perfect for storing powders, creams and liquids. They are made from high quality chemically resistant 316L stainless steel and are designed in such a way as to promote easy cleaning. The tri ferrule lid system means the containers can provide both an air and water tight seal.