SteriWare® GMP PP Tubs, White
Supplier: SAMPLING SYSTEMS LTD
8185P-500 8185P-250
77577-126EA 154.54 USD
77577-126 77577-124
SteriWare® GMP PP Tubs, White
Sample Containers General Purpose Containers
The polypropylene screw top containers are ideal for multiple applications including production, warehouse, sampling, and general laboratory work.
  • Robust polypropylene construction
  • Both EU and FDA compliant
  • Available in two volumes; 250 and 500 ml
  • Individually bagged and produced in a cleanroom
Tubs are manufactured in a cleanroom to ensure cleanliness and purity. The wide neck design makes them ideal for use with powders and creams.
Certificaciones: Containers conform to the latest standards including FDA, EU 10/2011, EC 1935/2004, and are BSE/TSE free.