SteriWare® Jugs, PP
Supplier: SAMPLING SYSTEMS LTD
8432P-1000 8432P-1000S 8432P-500 8432P-500S
77577-350EA 100.55 USD
77577-350 77577-352 77577-354 77577-356
SteriWare® Jugs, PP
Bottles Carboys
Made from PP (FDA, EC 1935/2004 and EU 10/2011 conforming).
  • Moulded and packed in a cleanroom
  • BSE/TSE free
  • Full batch traceability
  • Graduation marks at every 10 ml
These range of class leading jugs are available pre-sterilised (gamma irradiated). The ergonomic handle is rounded to make it comfortable to hold and use.
Envase: Individually bagged.