Mono Pocket Samplers
Supplier: SAMPLING SYSTEMS LTD
1430S-1000 1430S-1500 1430S-600
77576-258EA 715.55 USD
77576-258 77576-260 77576-262
Mono Pocket Samplers
Samplers Solid Samplers
The mono pocket sampler is designed for sampling free-flowing powders and granules from a single point.
  • Great for taking a single sample of free-flowing powders/granules
  • Available in a range of sizes
  • Can be easily stripped down for cleaning
  • Available from stock
  • Fast delivery
The mono pocket sampler is suitable for taking samples of free-flowing powders/granules from a single point. Available in various lengths, the GMP design ensures excellent durability for many applications.