SteriWare® Cake Cutters
Supplier: SAMPLING SYSTEMS LTD
8765H-4050 8765H-4050S
77577-512EA 99.51 USD
77577-512 77577-514
SteriWare® Cake Cutters
Samplers Solid Samplers
These circular cutters are perfect for taking samples of cake like material.
  • Made from FDA and EC compliant HDPE
  • 100% cleanroom manufactured and packed
  • Choice of sterile and non sterile
These individually bagged samplers are perfect for taking small 2 to 3 cm samples of cake-like material typically from in trays.