Removable Powder Sampling Tips, Stainless Steel
Supplier: SAMPLING SYSTEMS LTD
TIP-25P TIP-25R TIP-40R
77578-250EA 261.79 USD
77578-250 77578-252 77578-254
Removable Powder Sampling Tips, Stainless Steel
Samplers Solid Samplers
Removable tips can add additional effectiveness to powder sampling.
  • Optional range of tips for a variety of environments
  • Unscrew to enable thorough cleaning of the sampler
  • Available in varied diameters and styles
This range of sampling tips provide an excellent alternative to standard tips and can be chosen according to the environment in which the sampler is being used. They can all be unscrewed for easy cleaning and come in a variety of diameters and styles.