Powder Lances
Supplier: SAMPLING SYSTEMS LTD
2250A-1000 2250A-2000 2250A-600
77576-376EA 415.06 USD
77576-376 77576-378 77576-380
Powder Lances
Samplers Solid Samplers
The powder lance can be used to manually take large volume samples.
  • Cross sectional sampling without damage
  • Mostly used with cohesive powders
  • Horizontal use through a sack or elsewhere vertically
Suitable for use in the pharmaceutical industries.