SteriWare® Liquid Samplers
Supplier: SAMPLING SYSTEMS LTD
8051H-1100 8051H-1100S 8051H-450 8051H-450S 8051H-900 8051H-900S
77576-872EA 298.86 USD
77576-872 77576-874 77576-876 77576-878 77576-880 77576-882
SteriWare® Liquid Samplers
Samplers Fluid Samplers
These disposable Jumbo pipettes are ideal for taking samples from low viscosity liquids. Made from 100% virgin HDPE.
  • Quick and easy to use
  • Colour-coded tip indicates which end to open and hold
  • Cross-contamination risk is eliminated - can be used straight from the bag
  • No cleaning is required before or after sampling
  • Available as gamma irradiated sterile (SAL 10⁻⁶)
  • Suitable for use in ATEX zones 0, 1 and 2 (450 and 900 mm lengths only)
Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc).
Certificaciones: FDA acceptable grade conforms to CFR 177.1520.