Viscous Samplers
Supplier: SAMPLING SYSTEMS LTD
7000S-1000 7000S-1500 7000S-600
77576-584EA 796 USD
77576-584 77576-586 77576-588
Viscous Samplers
Samplers Fluid Samplers
These viscous samplers are designed for creams, shampoos, soaps, honey and other semi-viscous materials.
After sampling, the sampler can be completely dismantled, for a quick and thorough cleaning.