Instrument Trays
Supplier: SAMPLING SYSTEMS LTD
A674-215 A674-295 A674-395 A674-490
77578-096EA 21.92 USD
77578-096 77578-098 77578-100 77578-102
Instrument Trays
Trays/Dishes/Pots Instrument Trays/Dishes
Made from 304 stainless steel.
  • Good chemical resistance, very good temperature resistance
  • Crevice free design
  • High quality polished finish
Ideal for general laboratory use as well as for carrying surgical instruments, sampling tools etc.