SteriWare® Scrapers, Disposable
Supplier: SAMPLING SYSTEMS LTD
8785H-110 8785H-110S 8785H-75 8785H-75S
77577-520EA 273.55 USD
77577-520 77577-522 77577-524 77577-526
SteriWare® Scrapers, Disposable
Scrapers
These disposable scrapers are made from high quality HDPE. They are ideal in the manufacture of creams, ointments and pastes.
  • Moulded & packed in a cleanroom
  • Made from FDA and EU conforming HDPE
  • BSE/TSE-free
  • Available as gamma irradiated sterile (SAL 10-6)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
Full batch traceability
Envase: Individually bagged.