SteriWare® FoodScoop Scoops, Disposable
Supplier: SAMPLING SYSTEMS LTD
8045B-1000 8045B-1000S 8045B-2500 8045B-500
77576-750EA 152.67 USD
77576-750 77576-752 77576-760 77576-766
SteriWare® FoodScoop Scoops, Disposable
Scoops
These blue food scoops are ideal for use in the food industry.
  • Made from food grade Polypropylene
  • Available sterile and non sterile
  • Highly visible colour
  • Wide range of sizes
These blue food scoops can be easily spotted (if left behind in the powder) and are widely used throughout the food industry. They are both manufactured and packed in a cleanroom and are made from food grade polypropylene in line with HACCP guidelines.