SteriWare® Disposable Knives
Supplier: SAMPLING SYSTEMS LTD
8039A-100 8039H-100 8039H-100S
77576-716EA 36.7 USD
77576-716 77576-718 77576-720
SteriWare® Disposable Knives
Knives
These single use knives are made from food grade plastic (100% virgin HDPE). Ideal for sample preparation and general laboratory use.
  • Can be used to cut or split products
  • Available non sterile or gamma irradiated sterile (SAL 10⁻⁶)
  • Moulded and packed in a cleanroom
  • No cleaning is required before or after sampling
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • BSE/TSE-free
Certificaciones: Material conforms to FDA and EU 10/2011. Full batch traceability.