316L Stainless Steel Beakers
Supplier: SAMPLING SYSTEMS LTD
A736-2000 A736-3000 A736-500 A736-1000
77578-126EA 185.55 USD
77578-126 77578-128 77578-130 77578-124
316L Stainless Steel Beakers
Beakers
Beakers are designed for use with powders and liquids.
  • Ideal for use in the laboratory or production environment
  • Perfect for both powders and liquids
  • Available in convenient sizes
  • Available from stock
  • Fast delivery
This range of stainless steel beakers are made from chemically resistant 316L stainless steel, making them suitable for a wide range of the most demanding applications. Available in different sizes, they are a great choice for any work with both powder or liquid.