Graphene oxide 10mg/mL dispersion in water 10-30 µm
Supplier: TCI America
G0557-100ML
G0557-25ML
G0557-5ML
TCG0557-100MLEA
657
USD
TCG0557-100ML
TCG0557-25ML
TCG0557-5ML
Graphene oxide 10mg/mL dispersion in water 10-30 µm
Graphene oxide
Specification Test Results
Appearance | Red to Dark blue to Black clear liquid to cloudy liquid |
Concentration (with Evaporation residue) | 9.5 to 10.4 mg/mL |
pH | min. 2 (25 deg-C) |
Average size | 10 to 30 micro_m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...