1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Methoxy-4-[(4-pentylphenyl)ethynyl]benzene, 1-(4-Methoxyphenyl)-2-(4-pentylphenyl)acetylene, 1-(4-Methoxyphenyl)ethynyl-4-pentylbenzene

M3230-25G M3230-5G
TCM3230-25GEA 344 USD
TCM3230-25G TCM3230-5G
1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene ≥98.0% (by GC)
1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene
Formula: C₂₀H₂₂O
MW: 278.395 g/mol
Melting Pt: 45 °C
MDL Number: MFCD00187208
CAS Number: 39969-28-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR