Lead(II) acetate trihydrate ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

L0330-100G L0330-25G
TCL0330-100GEA 29 USD
TCL0330-100G TCL0330-25G
Lead(II) acetate trihydrate ≥99.0% (by titrimetric analysis)
Lead(II) acetate trihydrate
Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Boiling Pt: 280
Melting Pt: 75
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1 III
Merck Index: 13,05415

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR