LFM-A13 ≥98.0% (by HPLC)

Supplier: TCI America
Warning

L0309-10MG L0309-50MG
TCL0309-10MGEA 110 USD
TCL0309-10MG TCL0309-50MG
LFM-A13 ≥98.0% (by HPLC)
LFM-A13
Formula: C₁₁H₈Br₂N₂O₂
MW: 360.00 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02179204
CAS Number: 62004-35-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 7.30 to 8.20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR