trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥98.0% (by GC, titration analysis), purified by sublimation

Supplier: TCI America
Warning

Synonyms: (2E)-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, (E)-Ferulic acid, Trans-3-(4-hydroxy-3-methoxyphenyl)acrylic acid, trans-Ferulic acid

F1257-1G F1257-5G
TCF1257-1GEA 92 USD
TCF1257-1G TCF1257-5G
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥98.0% (by GC, titration analysis), purified by sublimation
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid
Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD00004400
CAS Number: 537-98-4
EINECS: 208-679-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR