(E)-6,6'-Dibromo-[3,3'-biindolinylidene]-2,2'-dione ≥96.0% (by HPLC, total nitrogen)

Supplier: TCI America

D5738-1G D5738-5G
TCD5738-1GEA 182 USD
TCD5738-1G TCD5738-5G
(E)-6,6'-Dibromo-[3,3'-biindolinylidene]-2,2'-dione ≥96.0% (by HPLC, total nitrogen)
(E)-6,6'-Dibromo-[3,3'-biindolinylidene]-2,2'-dione
Formula: C₁₆H₈Br₂N₂O₂
MW: 420.055 g/mol
MDL Number: MFCD29917500
CAS Number: 1351240-72-6

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR