4''-Ethyl-2'-fluoro-4-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4''-Ethyl-2'-fluoro-4-propyl-p-terphenyl

E1264-1G E1264-5G
TCE1264-1GEA 91 USD
TCE1264-1G TCE1264-5G
4''-Ethyl-2'-fluoro-4-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)
4''-Ethyl-2'-fluoro-4-propyl-1,1':4',1''-terphenyl
Formula: C₂₃H₂₃F
MW: 318.44 g/mol
MDL Number: MFCD01830216
CAS Number: 95759-44-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 69.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR