4-[(4-Isopropoxyphenyl)sulfonyl]phenol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Hydroxyphenyl 4-Isopropoxyphenyl sulphone, 4-((4-Isopropoxyphenyl)sulphonyl)phenol

I1045-25G I1045-5G
TCI1045-25GEA 204 USD
TCI1045-25G TCI1045-5G
4-[(4-Isopropoxyphenyl)sulfonyl]phenol ≥98.0% (by GC)
4-[(4-Isopropoxyphenyl)sulfonyl]phenol
Formula: C₁₅H₁₆O₄S
MW: 292.35 g/mol
MDL Number: MFCD01861244
CAS Number: 95235-30-6
EINECS: 405-520-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR