N-Methoxy-N-methylnicotinamide ≥96.0% (by GC)

Supplier: TCI America
Warning

M3099-1G M3099-5G
TCM3099-1GEA 96 USD
TCM3099-1G TCM3099-5G
N-Methoxy-N-methylnicotinamide ≥96.0% (by GC)
N-Methoxy-N-methylnicotinamide
Formula: C₈H₁₀N₂O₂
MDL Number: MFCD11847417
CAS Number: 95091-91-1

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR