JK-P3 ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3,4-Dimethoxy-N-(5-phenyl-1H-pyrazol-3-yl)benzamide

J0012-25MG J0012-5MG
TCJ0012-25MGEA 397 USD
TCJ0012-25MG TCJ0012-5MG
JK-P3 ≥98.0% (by HPLC, titration analysis)
JK-P3
Formula: C₁₈H₁₇N₃O₃
MW: 323.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22666405
CAS Number: 942655-44-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR