2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane ≥98.0% (by GC)

Supplier: TCI America
Warning

P2602-1G P2602-5G
TCP2602-1GEA 24 USD
TCP2602-1G TCP2602-5G
2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane ≥98.0% (by GC)
2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane
Formula: C₇H₁₅BO₂
MW: 142.01 g/mol
Boiling Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09027082
CAS Number: 94242-85-0
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR