3,5-Dibromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

D5670-1G D5670-200MG
TCD5670-1GEA 329 USD
TCD5670-1G TCD5670-200MG
3,5-Dibromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
3,5-Dibromo-5'-phenyl-1,1':3',1''-terphenyl
Formula: C₂₄H₁₆Br₂
MDL Number: MFCD32263474
CAS Number: 942132-66-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 145.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR