Ditrimethylolpropane tetraacrylate ≥75.0% (by GC) stabilized

Supplier: TCI America
Warning

Synonyms: [Oxybis(methylene)]bis(2-ethylpropane-3,2,1-triyl) tetraacrylate, Di(Trimethylolpropane)tetraacrylate, Acrylic acid [Oxybis(methylene)]bis(2-ethylpropane-3,2,1-triyl) tetraester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D5687-100G D5687-500G
TCD5687-100GEA 52 USD
TCD5687-100G TCD5687-500G
Ditrimethylolpropane tetraacrylate ≥75.0% (by GC) stabilized
Ditrimethylolpropane tetraacrylate
Formula: C₂₄H₃₄O₉
MW: 466.53 g/mol
MDL Number: MFCD00192118
CAS Number: 94108-97-1
EINECS: 302-434-9

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 75.0 %
Specific gravity (20/20) 1.1050 to 1.1150

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR