1-Ethynyl-4-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Nitrophenylacetylene

E1223-1G E1223-5G
TCE1223-1GEA 53 USD
TCE1223-1G TCE1223-5G
1-Ethynyl-4-nitrobenzene ≥98.0% (by GC)
1-Ethynyl-4-nitrobenzene
Formula: C₈H₅NO₂
MW: 147.13 g/mol
MDL Number: MFCD00024794
CAS Number: 937-31-5

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 149.0 to 153.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR