[(3-Triethoxysilyl)propyl]succinic anhydride ≥93.0% (by GC)

Supplier: TCI America
Warning

T3817-25G T3817-5G
TCT3817-25GEA 112.27 USD
TCT3817-25G TCT3817-5G
[(3-Triethoxysilyl)propyl]succinic anhydride ≥93.0% (by GC)
[(3-Triethoxysilyl)propyl]succinic anhydride
Formula: C₁₃H₂₄O₆Si
MDL Number: MFCD01320976
CAS Number: 93642-68-3
EINECS: 297-566-6

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 93.0 %
Purity (Morpholine Method) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR