5-Hydroxy-3-methyl-2(5H)-furanone ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Hydroxy-3-methyl-2,5-dihydrofuran-2-one, 5-Hydroxy-3-methylfuran-2(5H)-one

H1747-1G H1747-200MG
TCH1747-1GEA 195 USD
TCH1747-1G TCH1747-200MG
5-Hydroxy-3-methyl-2(5H)-furanone ≥95.0% (by GC)
5-Hydroxy-3-methyl-2(5H)-furanone
Formula: C₅H₆O₃
Boiling Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15145822
CAS Number: 931-23-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR