4,4,5,5-Tetramethyl-2-[10-(2-naphthyl)anthracen-9-yl]-1,3,2-dioxaborolane ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 10-(2-Naphthyl)anthracene-9-boronic acid Pinacol ester, 9-(2-Naphthyl)-10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracene

T3674-1G T3674-5G
TCT3674-1GEA 100 USD
TCT3674-1G TCT3674-5G
4,4,5,5-Tetramethyl-2-[10-(2-naphthyl)anthracen-9-yl]-1,3,2-dioxaborolane ≥97.0% (by GC)
4,4,5,5-Tetramethyl-2-[10-(2-naphthyl)anthracen-9-yl]-1,3,2-dioxaborolane
Formula: C₃₀H₂₇BO₂
MW: 430.35 g/mol
MDL Number: MFCD28978626
CAS Number: 922518-84-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR