1,4-Diaminobutane dihydroiodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Putrescine dihydroiodide, 1,4-Butanediamine dihydroiodide, Tetramethylenediamine dihydroiodide

D5686-1G D5686-5G
TCD5686-1GEA 46 USD
TCD5686-1G TCD5686-5G
1,4-Diaminobutane dihydroiodide ≥98.0% (by titrimetric analysis)
1,4-Diaminobutane dihydroiodide
Formula: C₄H₁₂N₂·₂HI
MW: 343.98 g/mol
CAS Number: 916849-52-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR