Potassium (Acetoxymethyl)trifluoroborate ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Potassium (acetoxymethyl)trifluoroborate

P2494-1G P2494-5G
TCP2494-1GEA 258 USD
TCP2494-1G TCP2494-5G
Potassium (Acetoxymethyl)trifluoroborate ≥96.0% (by titrimetric analysis)
Potassium (Acetoxymethyl)trifluoroborate
Formula: C₃H₅BF₃KO₂
MW: 179.97 g/mol
MDL Number: MFCD28023894
CAS Number: 910251-35-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR