Phenyl[3-(trifluoromethyl)phenyl]iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Phenyl[3-(trifluoromethyl)phenyl]iodonium triflate
P2413-1G
P2413-5G
TCP2413-1GEA
147
USD
TCP2413-1G
TCP2413-5G
Phenyl[3-(trifluoromethyl)phenyl]iodonium trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
Phenyl[3-(trifluoromethyl)phenyl]iodonium trifluoromethanesulfonate
Formula:
C₁₄H₉F₆IO₃S MW: 498.18 g/mol |
MDL Number:
MFCD21608481 CAS Number: 905718-46-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 95.0 % |
Melting point | 110.0 to 114.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...