alpha-Tocopherol polyethylene glycol succinate

Supplier: TCI America

Synonyms: Tocofersolan, Vitamin E Polyethylene Glycol Succinate, α-Tocopherol polyethylene glycol succinate

T3118-25G T3118-5G
TCT3118-25GEA 241 USD
TCT3118-25G TCT3118-5G
alpha-Tocopherol polyethylene glycol succinate
alpha-Tocopherol polyethylene glycol succinate
Formula: (C₂H₄O)nC₃₃H₅₄O₅
Melting Pt: 40 °C
CAS Number: 9002-96-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Solubility in Water almost transparency
Water max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR