4-(Naphthalen-2-yl)-N-phenylaniline ≥98.0% (by GC)

Supplier: TCI America

N1134-1G N1134-5G
TCN1134-1GEA 98 USD
TCN1134-1G TCN1134-5G
4-(Naphthalen-2-yl)-N-phenylaniline ≥98.0% (by GC)
4-(Naphthalen-2-yl)-N-phenylaniline
Formula: C₂₂H₁₇N
MDL Number: MFCD30478546
CAS Number: 897671-79-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 152.0 to 156.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR