N,9-Diphenyl-9H-carbazol-3-amine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 3-(N-Phenylamino)-9-phenyl-9H-carbazole

D5567-1G D5567-5G
TCD5567-1GEA 119 USD
TCD5567-1G TCD5567-5G
N,9-Diphenyl-9H-carbazol-3-amine ≥98.0% (by HPLC, total nitrogen)
N,9-Diphenyl-9H-carbazol-3-amine
Formula: C₂₄H₁₈N₂
MW: 334.42 g/mol
MDL Number: MFCD26401776
CAS Number: 894791-43-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR