Dolasetron Mesylate monohydrate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: rel-(5s,6R,8r,9aS)-3-Oxooctahydro-1H-2,6-methanoquinolizin-8-yl 1H-indole-3-carboxylate methanesulphonate hydrate, Dolasetron methanesulphonate, Dolasetron Mesylate hydrate

D5481-200MG D5481-50MG
TCD5481-200MGEA 471 USD
TCD5481-200MG TCD5481-50MG
Dolasetron Mesylate monohydrate ≥98.0% (by HPLC)
Dolasetron Mesylate monohydrate
Formula: C₁₉H₂₀N₂O₃·CH₄O₃S·H₂O
MW: 420.48 g/mol
MDL Number: MFCD01718979
CAS Number: 878143-33-0
Merck Index: 14,03411

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 6.10 to 6.70 %
Water 3.0 to 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR