1,5-Diacetyl-1,3,5-triazinane-2,4-dione ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D5634-1G D5634-5G
TCD5634-1GEA 83 USD
TCD5634-1G TCD5634-5G
1,5-Diacetyl-1,3,5-triazinane-2,4-dione ≥98.0% (by HPLC, titration analysis)
1,5-Diacetyl-1,3,5-triazinane-2,4-dione
Formula: C₇H₉N₃O₄
MW: 199.17 g/mol
MDL Number: MFCD00604438
CAS Number: 86320-44-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 212.0 to 216.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR