N,9,9-Triphenyl-9H-fluoren-2-amine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2-Phenylamino-9,9-diphenyl-9H-fluorene

T3746-1G T3746-5G
TCT3746-1GEA 147 USD
TCT3746-1G TCT3746-5G
N,9,9-Triphenyl-9H-fluoren-2-amine ≥98.0% (by HPLC, total nitrogen)
N,9,9-Triphenyl-9H-fluoren-2-amine
Formula: C₃₁H₂₃N
MW: 409.53 g/mol
MDL Number: MFCD30470208
CAS Number: 860465-14-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 206.0 to 210.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR